Thiophene-2-carbonyl chloride ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride

T0177-500G T0177-25G T0177-100G
TCT0177-500GEA 602 USD
TCT0177-500G TCT0177-25G TCT0177-100G
Thiophene-2-carbonyl chloride ≥99.0% (by titrimetric analysis)
Thiophene-2-carbonyl chloride

CAS Number: 5271-67-0
MDL Number: MFCD00005428
Molecular Formula: C5H3ClOS
Molecular Weight: 146.59
Purity/Analysis Method: >99.0% (T)
Form: Clear Liquid
Color: Very Pale Yellow
Boiling point (°C): 190
Flash Point (°C): 90
Specific Gravity (20/20): 1.38


Other Product Names / Synonyms: Thiophene-2-carbonyl Chloride

Formula: C₅H₃ClOS
MW: 146.6 g/mol
Boiling Pt: 190 °C
Density: 1.38 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD00005428
CAS Number: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5aac595de6b2d136dc1f23129b894f48157db4ae&vid=5aac595de6b2d136dc1f23129b894f48157db4ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR