Terephthalic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 100-21-0
MDL Number: MFCD00002558
Molecular Formula: C8H6O4
Molecular Weight: 166.13
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 402
Flash Point (°C): 300
Product Categories: Materials Chemistry, Metal Organic Framework (MOF) and Porous Coordination Polymer (PCP) Research Reagents, Monomers
Other Product Names / Synonyms: p-Phthalic Acid, Benzene-1,4-dicarboxylic Acid
Formula:
C₈H₆O₄ Boiling Pt: 392 °C (1013 hPa) Melting Pt: 402 °C Density: 1.472 g/cm³ (20 °C) Flash Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002558 CAS Number: 100-21-0 EINECS: 202-830-0 Merck Index: 12,09306 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...