Tetramethylsilane ≥99.0%

Supplier: TCI America
Danger

T0154-100ML T0154-25ML
TCT0154-100MLEA 120.04 USD
TCT0154-100ML TCT0154-25ML
Tetramethylsilane ≥99.0%
Tetramethylsilane

[for NMR]
CAS Number: 75-76-3
MDL Number: MFCD00008274
Molecular Formula: C4H12Si
Molecular Weight: 88.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 26
Flash Point (°C): -27
Specific Gravity (20/20): 0.64
Storage Temperature: 0-10°C

Formula: C₄H₁₂Si
Boiling Pt: 26 °C
Melting Pt: -99 °C
Density: 0.64 (20 °C)
Flash Pt: -27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008274
CAS Number: 75-76-3
EINECS: 200-899-1
UN: 2749
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2d6f43af8f59a29a1b1afb3e58cb6c0a220c5ea&vid=0d7f6557940b3beed579411bc7c31c67f2a11433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR