Tetramethylene sulphoxide ≥95.0%

Supplier: TCI America

Synonyms: Tetrahydrothiophene 1-oxide

T0146-25G T0146-500G
TCT0146-025GEA 71.1 USD
TCT0146-025G TCT0146-500G
Tetramethylene sulphoxide ≥95.0%
Tetramethylene sulphoxide

CAS Number: 1600-44-8
MDL Number: MFCD00005477
Molecular Formula: C4H8OS
Molecular Weight: 104.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 107
Flash Point (°C): 110
Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: Tetrahydrothiophene 1-Oxide

Formula: C₄H₈OS
Boiling Pt: 107 °C
Density: 1.18 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005477
CAS Number: 1600-44-8
EINECS: 216-493-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=284236c50699ea2e846ac7d5791172727ec9844b&vid=284236c50699ea2e846ac7d5791172727ec9844b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR