Titanium(IV) isopropoxide
Supplier: TCI America
T0133-500G
T0133-25G
TCT0133-500GEA
49.85
USD
TCT0133-500G
TCT0133-25G
Titanium(IV) isopropoxide
Titanium(IV) isopropoxide
CAS Number: 546-68-9
MDL Number: MFCD00008871
Molecular Formula: C12H28O4Ti
Molecular Weight: 284.22
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 220
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: Isopropyl Titanate, Tetraisopropyl Titanate, Titanium(IV) Isopropoxide, Titanium(IV) Tetraisopropoxide
Formula:
C₁₂H₂₈O₄Ti MW: 284.23 g/mol Boiling Pt: 220 °C Melting Pt: 18 - 20 °C Density: 0.97 (20 °C) Flash Pt: 51 °C |
MDL Number:
MFCD00008871 CAS Number: 546-68-9 EINECS: 208-909-6 UN: 2413 ADR: 3,III Merck Index: 12,09614 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...