Tetrahydropyran ≥98.0%

Supplier: TCI America

T0110-500ML T0110-25ML T0110-100ML
TCT0110-500MLEA 152.58 USD
TCT0110-500ML TCT0110-25ML TCT0110-100ML
Tetrahydropyran ≥98.0%
Tetrahydropyran

CAS Number: 142-68-7
MDL Number: MFCD00006585
Molecular Formula: C5H10O
Molecular Weight: 86.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 88
Flash Point (°C): -20
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: THP, Pentamethylene Oxide

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 88 °C
Melting Pt: –44.2 °C
Density: 0.88 (20 °C)
Flash Pt: –20 °C
MDL Number: MFCD00006585
CAS Number: 142-68-7
EINECS: 205-552-8
UN: 3271
ADR: 3,II
Merck Index: 12,09356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea5ab73fbe62acf59ab28320028c4ae0f55fd68&vid=5ea5ab73fbe62acf59ab28320028c4ae0f55fd68&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR