Tetrachloro-p-benzoquinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Tetrachloroquinone, 2,3,5,6-Tetrachloro-1,4-benzoquinone, 4-Chloranil, p-Chloranil, 2,3,5,6-Tetrachlorobenzo-1,4-quinone, Tetrachloro-1,4-benzoquinone

T0061-25G T0061-500G
TCT0061-025GEA 35.64 USD
TCT0061-025G TCT0061-500G
Tetrachloro-p-benzoquinone ≥98.0% (by GC, titration analysis)
Tetrachloro-p-benzoquinone

CAS Number: 118-75-2
MDL Number: MFCD00001594
Molecular Formula: C6Cl4O2
Molecular Weight: 245.86
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 297
Lambda max.: 510 nm (CH2Cl2)


Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: Spergon, Tetrachloro-p-benzoquinone

Formula: C₆Cl₄O₂
MW: 245.88 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 297 °C
Density: 1.97 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001594
CAS Number: 118-75-2
EINECS: 204-274-4
Merck Index: 13,02088
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=281ea4ae1f78d014fdf3e547a722e53f62829cc1&vid=281ea4ae1f78d014fdf3e547a722e53f62829cc1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR