Tetrabutylammonium iodide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
CAS Number: 311-28-4
MDL Number: MFCD00011636
Molecular Formula: C16H36IN
Molecular Weight: 369.38
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 147
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: TBAI
Formula:
C₁₆H₃₆IN Melting Pt: 147 °C Density: 1.0027 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011636 CAS Number: 311-28-4 EINECS: 206-220-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...