Terephthaloyl dichloride ≥99.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Terephthaloyl chloride
T0017-500G
T0017-25G
T0017-100G
TCT0017-500GEA
58
USD
TCT0017-500G
TCT0017-25G
TCT0017-100G
Terephthaloyl dichloride ≥99.0% (by GC, titration analysis)
Terephthaloyl dichloride
CAS Number: 100-20-9
MDL Number: MFCD00000693
Molecular Formula: C8H4Cl2O2
Molecular Weight: 203.02
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 266
Melting point (°C): 83
Flash Point (°C): 180
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Terephthaloyl Dichloride
Formula:
C₈H₄Cl₂O₂ MW: 203.02 g/mol Boiling Pt: 266 °C Melting Pt: 83 °C Density: 1.34 g/cm³ (100 °C) Flash Pt: 180 °C |
MDL Number:
MFCD00000693 CAS Number: 100-20-9 EINECS: 202-829-5 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...