n-Hexadecane

Supplier: TCI America
Danger

Synonyms: CetaneHexadecane

S0555-250ML
TCS0555-250MLEA 203 USD
TCS0555-250ML
n-Hexadecane
n-Hexadecane

Reference Material for Flash Point Certified by The Japan Petroleum Institute
CAS Number: 544-76-3
Molecular Formula: C16H34
Molecular Weight: 226.45
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: Hexadecane

Formula: H₃C(CH₂)₁₄CH₃
MW: 226.45 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.77331 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://pr.vwr.com/stibo/web/std.lang.all/41/85/28994185.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR