Sodium-tert-butanolate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate
S0450-100G
S0450-25G
TCS0450-100GEA
60.06
USD
TCS0450-100G
TCS0450-25G
Sodium-tert-butanolate ≥98.0% (by titrimetric analysis)
Sodium-tert-butanolate
CAS Number: 865-48-5
MDL Number: MFCD00040575
Molecular Formula: C4H9NaO
Molecular Weight: 96.10
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 270
Flash Point (°C): 14
Formula:
C₄H₉NaO MW: 96.1 g/mol Melting Pt: 270 °C Flash Pt: 14 °C |
MDL Number:
MFCD00040575 CAS Number: 865-48-5 EINECS: 212-741-9 UN: 3206 ADR: 4.2,II |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...