4-Sulfamoylbenzoic acid ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Sulphamoylbenzoic acid

S0323-500G S0323-25G
TCS0323-500GEA 296 USD
TCS0323-500G TCS0323-025G
4-Sulfamoylbenzoic acid ≥95.0% (by titrimetric analysis)
4-Sulphamoylbenzoic acid

CAS Number: 138-41-0
MDL Number: MFCD00007938
Molecular Formula: C7H7NO4S
Molecular Weight: 201.20
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-Carboxybenzenesulfonamide, Carzenide

Formula: C₇H₇NO₄S
MW: 201.2 g/mol
MDL Number: MFCD00007938
CAS Number: 138-41-0
EINECS: 205-327-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6b57a5cef86dbd82be0b3a0371504dd54bcbacf&vid=a6b57a5cef86dbd82be0b3a0371504dd54bcbacf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR