Succinonitrile ≥99.0%

Supplier: TCI America
Warning

Synonyms: Butanedinitrile, Succinodinitrile

S0109-25G S0109-500G
TCS0109-025GEA 31.3 USD
TCS0109-025G TCS0109-500G
Succinonitrile ≥99.0%
Succinonitrile

CAS Number: 110-61-2
MDL Number: MFCD00001949
Molecular Formula: C4H4N2
Molecular Weight: 80.09
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 159
Flash Point (°C): 132
Freezing point (°C): 57


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Butanedinitrile, 1,2-Dicyanoethane, Ethylene Cyanide, Ethylene Dicyanide

Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 159 °C
Melting Pt: 53 °C
Density: 1.023 g/cm³ (20 °C)
Flash Pt: 132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001949
CAS Number: 110-61-2
EINECS: 203-783-9
UN: 3439
ADR: 6.1,III
Merck Index: 12,09041
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb58672f380c9566c68fd21303fef9857f6d88fb&vid=bb58672f380c9566c68fd21303fef9857f6d88fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR