Styrene ≥99.0% stabilized

Supplier: TCI America
Danger

Synonyms: Phenylethene, Phenylethylene, Vinylbenzene

Stabilization: Stabilised with Stabilised

S0095-500ML S0095-25ML
TCS0095-500MLEA 43.87 USD
TCS0095-500ML TCS0095-25ML
Styrene ≥99.0% stabilized
Styrene

(stabilized with TBC)
CAS Number: 100-42-5
MDL Number: MFCD00008612
Molecular Formula: C8H8
Molecular Weight: 104.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Melting point (°C): -31
Flash Point (°C): 32
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Styrol, Vinylbenzene

Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 145 °C
Melting Pt: -31 °C
Density: 0.91 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00008612
CAS Number: 100-42-5
EINECS: 202-851-5
UN: 2055
ADR: 3,III
Merck Index: 13,08944
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f73ab98d681693e869739e38317e23d32abe467&vid=2f73ab98d681693e869739e38317e23d32abe467&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR