Potassium sorbate ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

S0057-25G S0057-500G
TCS0057-025GEA 22.49 USD
TCS0057-025G TCS0057-500G
Potassium sorbate ≥99.0% (by titrimetric analysis)
Potassium sorbate

CAS Number: 24634-61-5
MDL Number: MFCD00016546
Molecular Formula: C6H8O2
Molecular Weight: 150.22
Purity/Analysis Method: >99.0% (T)
Form: Granul
Color: White


Other Product Names / Synonyms: Potassium 2,4-Hexadienoate, Sorbic Acid Potassium Salt

Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: 270 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016546
CAS Number: 24634-61-5
EINECS: 246-376-1
Merck Index: 13,07756
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76ba039900dd4825054e19c4290f3c8f545693ef&vid=76ba039900dd4825054e19c4290f3c8f545693ef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR