Ethyl sorbate ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl trans,trans-hexa-2,4-dienoate, (2E,4E)-Ethyl hexa-2,4-dienoate, Ethyl (2E,4E)-hexa-2,4-dienoate, 2,4-Hexadienoic acid ethyl ester, Sorbic acid ethyl ester, (2E,4E)-2,4-Hexadienoic acid ethyl ester, trans,trans-2,4-Hexadienoic acid ethyl ester, (E,E)-2,4-Hexadienoic acid ethyl ester, Ethyl (E,E)-hexa-2,4-dienoate
CAS Number: 2396-84-1
MDL Number: MFCD00009296
Molecular Formula: C8H12O2
Molecular Weight: 140.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 76
Flash Point (°C): 86
Specific Gravity (20/20): 0.94
Other Product Names / Synonyms: Sorbic Acid Ethyl Ester
Formula:
C₈H₁₂O₂ Boiling Pt: 76 °C Density: 0.94 (20 °C) Flash Pt: 86 °C |
MDL Number:
MFCD00009296 CAS Number: 2396-84-1 EINECS: 219-258-2 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...