Semicarbazide hydrochloride ≥99.0%

Supplier: TCI America
Danger

Synonyms: Semicarbazide hydrochloride, Carbamylhydrazine hydrochloride, Aminourea hydrochloride, Hydrazinecarboxamide hydrochloride, N-Aminourea hydrochloride, Hydrazinecarboxamidehydrochloride

S0032-500G S0032-25G
TCS0032-500GEA 78.32 USD
TCS0032-500G TCS0032-25G
Semicarbazide hydrochloride ≥99.0%
Semicarbazide hydrochloride

CAS Number: 563-41-7
MDL Number: MFCD00013009
Molecular Formula: CH5N3O
Molecular Weight: 111.53
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Color: White

Formula: CH₆ClN₃O
MW: 111.53 g/mol
Melting Pt: 174…178 °C
MDL Number: MFCD00013009
CAS Number: 563-41-7
EINECS: 209-247-0
UN: 2811
ADR: 6.1 III
Merck Index: 12,08588
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d782b35d8db77b8d65f90581338eaa81537c1c32&vid=d782b35d8db77b8d65f90581338eaa81537c1c32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR