Sebaconitrile ≥98.0%

Supplier: TCI America

Synonyms: 1,8-Dicyanooctane, Sebacic acid dinitrile

S0029-500ML S0029-25ML
TCS0029-500MLEA 523 USD
TCS0029-500ML TCS0029-25ML
Sebaconitrile ≥98.0%
Sebaconitrile

CAS Number: 1871-96-1
MDL Number: MFCD00019900
Molecular Formula: C10H16N2
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 200
Flash Point (°C): 110
Freezing point (°C): 7
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Decanedinitrile, 1,8-Dicyanooctane, Octamethylene Dicyanide

Formula: C₁₀H₁₆N₂
MW: 164.25 g/mol
Boiling Pt: 200 °C
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00019900
CAS Number: 1871-96-1
EINECS: 217-494-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f68d3c31338d8c6d33a82f3490b9fb25519dd197&vid=f68d3c31338d8c6d33a82f3490b9fb25519dd197&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR