Methyl salicylate ≥99.0%

Supplier: TCI America
Warning

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

S0015-500G S0015-25G
TCS0015-500GEA 42.35 USD
TCS0015-500G TCS0015-25G
Methyl salicylate ≥99.0%
Methyl salicylate

CAS Number: 119-36-8
MDL Number: MFCD00002214
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 224
Melting point (°C): -9
Flash Point (°C): 97
Specific Gravity (20/20): 1.19


Product Categories: Bioscience, Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 2-Hydroxybenzoic Acid Methyl Ester, Salicylic Acid Methyl Ester, Methyl 2-Hydroxybenzoate

Formula: 2-(HO)C₆H₄CO₂CH₃
MW: 152.15 g/mol
Boiling Pt: 432
Melting Pt: –9 °C
Density: 1.19 (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e9d68e443778019f895096cc9c89cc169c0b45d&vid=1e9d68e443778019f895096cc9c89cc169c0b45d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR