Ethyl salicylate ≥99.0%
Supplier: TCI America
|
Warning
|
CAS Number: 118-61-6
MDL Number: MFCD00002215
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 231
Melting point (°C): 1
Flash Point (°C): 120
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Ethyl 2-Hydroxybenzoate, 2-Hydroxybenzoic Acid Ethyl Ester, Salicylic Acid Ethyl Ester
Formula:
C₉H₁₀O₃ Boiling Pt: 231 °C Melting Pt: 1 °C Density: 1.13 (20 °C) Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002215 CAS Number: 118-61-6 EINECS: 204-265-5 Merck Index: 13,03881 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...