Resorcinol ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1,3-Dihydroxybenzene, 1,3-Benzenediol
R0008-500G
R0008-25G
TCR0008-500GEA
78.33
USD
TCR0008-500G
TCR0008-25G
Resorcinol ≥99.0%
Resorcinol
CAS Number: 108-46-3
MDL Number: MFCD00002269
Molecular Formula: C6H6O2
Molecular Weight: 110.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 178
Melting point (°C): 111
Flash Point (°C): 164
Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: 1,3-Dihydroxybenzene
Formula:
C₆H₆O₂ MW: 110.11 g/mol Boiling Pt: 178 °C Melting Pt: 111 °C Density: 1.272 g/cm³ (20 °C) Flash Pt: 164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002269 CAS Number: 108-46-3 EINECS: 203-585-2 UN: 2876 ADR: 6.1,III Merck Index: 12,08323 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...