Quinhydrone

Supplier: TCI America
Danger

Synonyms: Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)

Q0008-500G Q0008-25G Q0008-100G
TCQ0008-500GEA 307.6 USD
TCQ0008-500G TCQ0008-25G TCQ0008-100G
Quinhydrone
Quinhydrone

CAS Number: 106-34-3
MDL Number: MFCD00010310
Molecular Formula: C12H10O4
Molecular Weight: 218.21
Form: Crystal
Color: Yellow
Melting point (°C): 172

Formula: C₆H₄(OH)₂·C₆H₄O₂
MW: 218.21 g/mol
Melting Pt: 172 °C
Density: 1.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010310
CAS Number: 106-34-3
EINECS: 203-387-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,08148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b51e6f5d10be3401860232c0c920c0ce2b215225&vid=b51e6f5d10be3401860232c0c920c0ce2b215225&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR