p-Quaterphenyl ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
1,1':4',1'':4'',1'''-Quaterphenyl
Q0001-1G
Q0001-25G
Q0001-5G
TCQ0001-001GEA
59
USD
TCQ0001-001G
TCQ0001-25G
TCQ0001-005G
p-Quaterphenyl ≥98.0% (by HPLC)
p-Quaterphenyl
CAS Number: 135-70-6
MDL Number: MFCD00003062
Molecular Formula: C24H18
Molecular Weight: 306.41
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Formula:
C₂₄H₁₈ MW: 306.41 g/mol Boiling Pt: 517 °C (1013 hPa) Melting Pt: 308 °C |
MDL Number:
MFCD00003062 CAS Number: 135-70-6 EINECS: 205-213-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...