1,2,3,4,5-Pentamethylcyclopentadiene ≥93.0%

Supplier: TCI America

P1292-5ML P1292-25ML
TCP1292-005MLEA 137 USD
TCP1292-005ML TCP1292-025ML
1,2,3,4,5-Pentamethylcyclopentadiene ≥93.0%
1,2,3,4,5-Pentamethylcyclopentadiene

CAS Number: 4045-44-7
MDL Number: MFCD00001354
Molecular Formula: C10H16
Molecular Weight: 136.24
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 60
Flash Point (°C): 44
Specific Gravity (20/20): 0.84

Formula: C₁₀H₁₆
Boiling Pt: 60 °C
Density: 0.84 (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00001354
CAS Number: 4045-44-7
EINECS: 223-743-4
UN: 3295
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dac43f66c73c5e376d452ac4b548111d43bee0c&vid=9dac43f66c73c5e376d452ac4b548111d43bee0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR