1,1-Dimethoxyacetone ≥97.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
P1264-25ML
P1264-100ML
TCP1264-25MLEA
76.32
USD
TCP1264-25ML
TCP1264-100ML
1,1-Dimethoxyacetone ≥97.0%
1,1-Dimethoxyacetone
CAS Number: 6342-56-9
MDL Number: MFCD00008758
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 141
Flash Point (°C): 37
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 1,1-Dimethoxyacetone, 1,1-Dimethoxy-2-propanone, Methylglyoxal Dimethyl Acetal
Formula:
C₅H₁₀O₃ MW: 118.13 g/mol Boiling Pt: 141 °C Melting Pt: –57 °C Density: 1.00 (20 °C) Flash Pt: 37 °C |
MDL Number:
MFCD00008758 CAS Number: 6342-56-9 EINECS: 228-735-4 UN: 1224 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...