2-Phenoxybenzoic acid
Supplier: TCI America
|
Warning
|
P1252-25G
P1252-5G
TCP1252-025GEA
195
USD
TCP1252-025G
TCP1252-5G
2-Phenoxybenzoic acid
2-Phenoxybenzoic acid
CAS Number: 2243-42-7
MDL Number: MFCD00002429
Molecular Formula: C13H10O3
Molecular Weight: 214.22
Form: Crystal
Color: White
Boiling point (°C): 355
Melting point (°C): 113
Formula:
C₁₃H₁₀O₃ MW: 214.22 g/mol Boiling Pt: 355 °C Melting Pt: 113 °C Density: 1.2141 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002429 CAS Number: 2243-42-7 EINECS: 218-811-5 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 112.0 to 115.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...