Propiolaldehyde diethyl acetal ≥97.0%
Supplier: TCI America
P1173-25ML
P1173-5ML
TCP1173-025MLEA
352
USD
TCP1173-025ML
TCP1173-005ML
Propiolaldehyde diethyl acetal ≥97.0%
Propiolaldehyde diethyl acetal
CAS Number: 10160-87-9
MDL Number: MFCD00009237
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 139
Flash Point (°C): 32
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 3,3-Diethoxy-1-propyne, Propiolaldehyde Diethyl Acetal
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 139 °C Density: 0.89 (20 °C) Flash Pt: 32 °C |
MDL Number:
MFCD00009237 CAS Number: 10160-87-9 EINECS: 233-430-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...