L-Phenylalaninol ≥98.0%

Supplier: TCI America

Synonyms: (S)-(-)-2-Amino-3-phenyl-1-propanol, (S)-(-)-Phenylalaninol, (S)-2-Amino-3-phenylpropan-1-ol, L(-)-2-Amino-3-phenyl-1-propanol, L(-)-Phenylalaninol, H-Phenylalaninol, L(-)-2-Amino-3-phenylpropan-1-ol, (S)-(-)-2-Amino-3-phenylpropan-1-ol

P1028-25G P1028-5G
TCP1028-025GEA 185 USD
TCP1028-025G TCP1028-005G
L-Phenylalaninol ≥98.0%
L-Phenylalaninol

CAS Number: 3182-95-4
MDL Number: MFCD00004732
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 94
Specific rotation [a]20/D: -24.5 deg (C=5, EtOH)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Amino-3-phenyl-1-propanol

Formula: C₉H₁₃NO
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 94 °C
MDL Number: MFCD00004732
CAS Number: 3182-95-4
EINECS: 221-674-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92577bfba10a2bc4b728c6a3944dcef5df349e39&vid=92577bfba10a2bc4b728c6a3944dcef5df349e39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR