Potassium hexafluorophosphate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
P1023-500G
P1023-25G
TCP1023-500GEA
193.52
USD
TCP1023-500G
TCP1023-25G
Potassium hexafluorophosphate ≥95.0% (by titrimetric analysis)
Potassium hexafluorophosphate
CAS Number: 17084-13-8
MDL Number: MFCD00011412
Molecular Formula: KPF6
Molecular Weight: 184.06
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White
Melting point (°C): 575
Formula:
F₆KP Melting Pt: 575 °C Density: 2.58 g/cm³ (20 °C) |
MDL Number:
MFCD00011412 CAS Number: 17084-13-8 EINECS: 241-143-0 UN: 3260 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...