Phenyl vinyl sulphone ≥98.0%

Supplier: TCI America

Synonyms: Vinyl phenyl sulphone, (Vinylsulphonyl)benzene

P0982-25G P0982-5G
TCP0982-25GEA 215 USD
TCP0982-25G TCP0982-005G
Phenyl vinyl sulphone ≥98.0%
Phenyl vinyl sulphone

CAS Number: 5535-48-8
MDL Number: MFCD00007554
Molecular Formula: C8H8O2S
Molecular Weight: 168.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 68

Formula: C₈H₈O₂S
Melting Pt: 68 °C
MDL Number: MFCD00007554
CAS Number: 5535-48-8
EINECS: 226-890-2

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR