4-Phenylcyclohexanone ≥98.0%
Supplier: TCI America
Synonyms:
4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane
P0975-25G
P0975-250G
TCP0975-025GEA
129.7
USD
TCP0975-025G
TCP0975-250G
4-Phenylcyclohexanone ≥98.0%
4-Phenylcyclohexanone
CAS Number: 4894-75-1
MDL Number: MFCD00001641
Molecular Formula: C12H14O
Molecular Weight: 174.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 78
Formula:
C₁₂H₁₄O Boiling Pt: 294 °C (1013 hPa) Melting Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001641 CAS Number: 4894-75-1 EINECS: 225-517-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...