Pyridinium dichromate ≥98.0%

Supplier: TCI America

Synonyms: PDC

P0931-500G P0931-25G P0931-100G
TCP0931-500GEA 156.7 USD
TCP0931-500G TCP0931-25G TCP0931-100G
Pyridinium dichromate ≥98.0%
Pyridinium dichromate

CAS Number: 20039-37-6
MDL Number: MFCD00013105
Molecular Formula: C10H12Cr2N2O7
Molecular Weight: 376.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Reddish Yellow
Melting point (°C): 149


Product Categories: Synthetic Organic Chemistry, Oxidation
Other Product Names / Synonyms: PDC, Cornforth Reagent

Formula: C₁₀N₁₂Cr₂N₂O₇
Melting Pt: 149 °C
MDL Number: MFCD00013105
CAS Number: 20039-37-6
EINECS: 243-478-8
UN: 3087
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c02e6bd7741b03beea7c07f0aed56ca0374f163&vid=6c02e6bd7741b03beea7c07f0aed56ca0374f163&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR