N,N,N-Trimethylanilinium tribromide ≥98.0%

Supplier: TCI America

Synonyms: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

P0928-500G P0928-25G
TCP0928-500GEA 337 USD
TCP0928-500G TCP0928-25G
N,N,N-Trimethylanilinium tribromide ≥98.0%
N,N,N-Trimethylanilinium tribromide

CAS Number: 4207-56-1
MDL Number: MFCD00011789
Molecular Formula: C9H14Br3N
Molecular Weight: 375.93
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 115


Product Categories: Synthetic Organic Chemistry, Halogenation
Other Product Names / Synonyms: Phenyltrimethylammonium Tribromide

Formula: C₉H₁₄Br₃N
Melting Pt: 115 °C
MDL Number: MFCD00011789
CAS Number: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2659e61e7f3f02cbb474d3fdf3c29e7a706f3d9&vid=f2659e61e7f3f02cbb474d3fdf3c29e7a706f3d9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR