N-Phenylmaleimide ≥98.0%

Supplier: TCI America

P0900-25G P0900-100G
TCP0900-25GEA 44.79 USD
TCP0900-25G TCP0900-100G
N-Phenylmaleimide ≥98.0%
N-Phenylmaleimide

CAS Number: 941-69-5
MDL Number: MFCD00005502
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: >98.0% (GC)
Form: Flake
Color: Yellow
Boiling point (°C): 181
Melting point (°C): 90
Flash Point (°C): 176

Formula: C₁₀H₇NO₂
Boiling Pt: 181 °C
Melting Pt: 90 °C
Flash Pt: 176 °C
MDL Number: MFCD00005502
CAS Number: 941-69-5
EINECS: 213-382-0
UN: 2811
ADR: 6.1,III
Merck Index: 12,07452
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=654e129a010772bcfc89430700fb619f5c7981b3&vid=654e129a010772bcfc89430700fb619f5c7981b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR