N,N,N',N',N''-Pentamethyldiethylenetriamine ≥99.0%
Supplier: TCI America
Synonyms:
N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine
P0881-500ML
P0881-25ML
TCP0881-500MLEA
106.53
USD
TCP0881-500ML
TCP0881-25ML
N,N,N',N',N''-Pentamethyldiethylenetriamine ≥99.0%
N,N,N',N',N''-Pentamethyldiethylenetriamine
CAS Number: 3030-47-5
MDL Number: MFCD00014876
Molecular Formula: C9H23N3
Molecular Weight: 173.30
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 86
Melting point (°C): -20
Flash Point (°C): 72
Specific Gravity (20/20): 0.83
Product Categories: Materials Chemistry, Precise Polymerization Reagents, Polymerization Catalysts
Other Product Names / Synonyms: PMDTA, PMDETA
Formula:
C₉H₂₃N₃ MW: 173.30 g/mol Boiling Pt: 86 °C Melting Pt: -20 °C Density: 0.83 (20 °C) Flash Pt: 72 °C |
MDL Number:
MFCD00014876 CAS Number: 3030-47-5 EINECS: 221-201-1 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...