Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥95.0%

Supplier: TCI America

Synonyms: (Chloro)(perfluorophenyl)dimethylsilane

P0854-1ML P0854-5ML
TCP0854-001MLEA 114 USD
TCP0854-001ML TCP0854-5ML
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥95.0%
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride)

[Pentafluorophenyldimethylsilylating Agent]
CAS Number: 20082-71-7
MDL Number: MFCD00000500
Molecular Formula: C8H6ClF5Si
Molecular Weight: 260.66
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 47
Flash Point (°C): 75
Specific Gravity (20/20): 1.41


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Silane Coupling Agents, Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: Dimethylpentafluorophenylchlorosilane, Flophemesyl Chloride

Formula: C₈H₆ClF₅Si
MW: 260.67 g/mol
Boiling Pt: 47 °C
Density: 1.41 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00000500
CAS Number: 20082-71-7
EINECS: 243-506-9
UN: 2987
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee3dd4b6500331aef747888b2c9a0f929922de96&vid=55867f7b9476ed87c59f2688740ae332c38a9ea6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR