3-Phenoxytoluene ≥98.0%

Supplier: TCI America

P0831-25ML P0831-500ML
TCP0831-025MLEA 51 USD
TCP0831-025ML TCP0831-500ML
3-Phenoxytoluene ≥98.0%
3-Phenoxytoluene

CAS Number: 3586-14-9
MDL Number: MFCD00008531
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 272
Flash Point (°C): 113
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 3-Methyldiphenyl Ether, m-Tolyl Phenyl Ether

Formula: C₁₃H₁₂O
Boiling Pt: 272 °C
Density: 1.05 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008531
CAS Number: 3586-14-9
EINECS: 222-716-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8f7374c53d78f8d579c57c87497d299bfca1f2f&vid=b8f7374c53d78f8d579c57c87497d299bfca1f2f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR