(R)-(+)-α-Methylbenzylamine ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
(R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine
CAS Number: 3886-69-9
MDL Number: MFCD00064405
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 70
Specific Gravity (20/20): 0.95
Specific rotation [a]20/D: 39 deg (neat)
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-(+)-alpha-Methylbenzylamine
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 85 °C Melting Pt: 32.5 °C Density: 0.95 (20 °C) Flash Pt: 70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064405 CAS Number: 3886-69-9 EINECS: 223-423-4 Merck Index: 12,06106 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...