(S)-(-)-α-Methylbenzylamine ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
(S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine
CAS Number: 2627-86-3
MDL Number: MFCD00064406
Molecular Formula: C8H11N
Molecular Weight: 121.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 84
Flash Point (°C): 79
Specific Gravity (20/20): 0.95
Specific rotation [a]20/D: -39 deg (neat)
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S)-(-)-alpha-Methylbenzylamine
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 84 °C Melting Pt: -10 °C Density: 0.95 (20 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064406 CAS Number: 2627-86-3 EINECS: 220-098-0 Merck Index: 12,06106 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...