4-(Aminomethyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Picolamine, (4-Pyridyl)methylamine, Pyridin-4-ylmethanamine, 4-Picolylamine, 4-Pyridylmethylamine

P0779-500G P0779-25G P0779-100G
TCP0779-500GEA 588 USD
TCP0779-500G TCP0779-25G TCP0779-100G
4-(Aminomethyl)pyridine ≥98.0% (by GC, titration analysis)
4-(Aminomethyl)pyridine

CAS Number: 3731-53-1
MDL Number: MFCD00006449
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 230
Melting point (°C): -8
Flash Point (°C): 104
Specific Gravity (20/20): 1.07


Other Product Names / Synonyms: 4-(Aminomethyl)pyridine

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 230 °C
Melting Pt: –8 °C
Density: 1.07 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00006449
CAS Number: 3731-53-1
EINECS: 223-092-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b71ab502590cad23c7acf1df0cc834747ad2e6d0&vid=b71ab502590cad23c7acf1df0cc834747ad2e6d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR