3-(Aminomethyl)pyridine ≥99.0%
Supplier: TCI America
Synonyms:
3-Pyridylmethylamine, (3-Pyridyl)methylamine, 3-Picolamine, Pyridin-3-ylmethanamine, 3-Picolylamine
P0762-500ML
P0762-25ML
TCP0762-500MLEA
172.9
USD
TCP0762-500ML
TCP0762-25ML
3-(Aminomethyl)pyridine ≥99.0%
3-(Aminomethyl)pyridine
CAS Number: 3731-52-0
MDL Number: MFCD00006412
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Melting point (°C): -21
Flash Point (°C): 100
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 3-(Aminomethyl)pyridine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 74 °C Melting Pt: –21 °C Density: 1.07 (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00006412 CAS Number: 3731-52-0 EINECS: 223-091-0 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...