Phenolphthalein 0.1% in ethanol 90%
Supplier: TCI America
|
Danger
|
P0701-100ML
P0701-500ML
TCP0701-100MLEA
26
USD
TCP0701-100ML
TCP0701-500ML
Phenolphthalein 0.1% in ethanol 90%
Phenolphthalein
[for Titration]
CAS Number: 77-09-8
MDL Number: MFCD00005913
Molecular Weight: 318.33
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 13
Formula:
C₂₀H₁₄O₄ Flash Pt: 13 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...