2-Phenylpropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 492-37-5
MDL Number: MFCD00002650
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 267
Melting point (°C): -5
Flash Point (°C): 148
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: Hydratropic Acid
Formula:
C₉H₁₀O₂ Boiling Pt: 267 °C Melting Pt: –5 °C Density: 1.10 (20 °C) Flash Pt: 148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002650 CAS Number: 492-37-5 EINECS: 207-752-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...