4-Phenylbutyric acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Phenylbutanoic acid
P0643-250G
P0643-25G
TCP0643-250GEA
310.7
USD
TCP0643-250G
TCP0643-025G
4-Phenylbutyric acid ≥98.0% (by GC, titration analysis)
4-Phenylbutyric acid
CAS Number: 1821-12-1
MDL Number: MFCD00004403
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 175
Melting point (°C): 52
Flash Point (°C): 100
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 175 °C Melting Pt: 52 °C Flash Pt: 100 °C |
MDL Number:
MFCD00004403 CAS Number: 1821-12-1 EINECS: 217-341-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...