Diisopropyl phosphonate ≥98.0%
Supplier: TCI America
Synonyms:
Diisopropyl phosphite, Di-i-propylphosphite, Diisopropyl hydrogen phosphite
CAS Number: 1809-20-7
MDL Number: MFCD00117905
Molecular Formula: C6H15O3P
Molecular Weight: 166.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 112
Specific Gravity (20/20): 1.00
Product Categories: Synthetic Organic Chemistry, Dialkyl Phosphites (Multi-component Reaction)
Other Product Names / Synonyms: Diisopropyl Phosphonate, Phosphonic Acid Diisopropyl Ester, Phosphorous Acid Diisopropyl Ester
Formula:
C₆H₁₅O₃P MW: 166.16 g/mol Boiling Pt: 79 °C Density: 1.00 (20 °C) Flash Pt: 112 °C |
MDL Number:
MFCD00117905 CAS Number: 1809-20-7 EINECS: 217-317-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...