Pyruvic acid ≥97.0%

Supplier: TCI America
Danger

Synonyms: α-Ketopropionic acid, 2-Oxopropionic acid, 2-Ketopropionic acid, 2-Oxopropanoic acid

P0579-500G P0579-25G
TCP0579-500GEA 159 USD
TCP0579-500G TCP0579-25G
Pyruvic acid ≥97.0%
Pyruvic acid

CAS Number: 127-17-3
MDL Number: MFCD00002585
Molecular Formula: C3H4O3
Molecular Weight: 88.06
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Melting point (°C): 13
Flash Point (°C): 82
Specific Gravity (20/20): 1.27
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Ketopropionic Acid, 2-Oxopropionic Acid

Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 62 °C
Melting Pt: 13 °C
Density: 1.27 (20 °C)
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002585
CAS Number: 127-17-3
EINECS: 204-824-3
UN: 3265
ADR: 8,II
Merck Index: 12,08205
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a25c351d6b1db9090e355c94537efe6f40d39f72&vid=a25c351d6b1db9090e355c94537efe6f40d39f72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR