Pyrocatechol ≥99.0%

Supplier: TCI America
Warning

Synonyms: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene

P0567-500G P0567-25G
TCP0567-500GEA 85.02 USD
TCP0567-500G TCP0567-25G
Pyrocatechol ≥99.0%
Pyrocatechol

CAS Number: 120-80-9
MDL Number: MFCD00002188
Molecular Formula: C6H6O2
Molecular Weight: 110.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 246
Melting point (°C): 105
Flash Point (°C): 127


Other Product Names / Synonyms: 1,2-Dihydroxybenzene, Pyrocatechol

Formula: C₆H₆O₂
MW: 110.11 g/mol
Boiling Pt: 246 °C
Melting Pt: 105 °C
Density: 1.0548 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002188
CAS Number: 120-80-9
EINECS: 204-427-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,08089
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11d930726c5066d1c47c5723e8e927d174ad8ea8&vid=11d930726c5066d1c47c5723e8e927d174ad8ea8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR