3-Pyridylmethanol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol

P0555-500ML P0555-25ML
TCP0555-500MLEA 413.2 USD
TCP0555-500ML TCP0555-025ML
3-Pyridylmethanol ≥98.0%
3-Pyridylmethanol

CAS Number: 100-55-0
MDL Number: MFCD00006407
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 157
Specific Gravity (20/20): 1.14


Other Product Names / Synonyms: 3-(Hydroxymethyl)pyridine, Nicotinyl Alcohol

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 145 °C
Melting Pt: –7 °C
Density: 1.14 (20 °C)
Flash Pt: 157 °C
MDL Number: MFCD00006407
CAS Number: 100-55-0
EINECS: 202-864-6
Merck Index: 12,06614
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac4814d0293bc015ae762accca3949c5057e6f5a&vid=ac4814d0293bc015ae762accca3949c5057e6f5a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR