Dipropyl ether ≥98.0%

Supplier: TCI America
Danger

P0527-500ML P0527-25ML
TCP0527-500MLEA 696 USD
TCP0527-500ML TCP0527-25ML
Dipropyl ether ≥98.0%
Dipropyl ether

CAS Number: 111-43-3
MDL Number: MFCD00009376
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Melting point (°C): -123
Flash Point (°C): 21
Specific Gravity (20/20): 0.75


Other Product Names / Synonyms: Dipropyl Ether

Formula: C₆H₁₄O
Boiling Pt: 90 °C
Melting Pt: –123 °C
Density: 0.75 (20 °C)
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00009376
CAS Number: 111-43-3
EINECS: 203-869-6
UN: 2384
ADR: 3,II
Merck Index: 12,08042
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78ea7a3679ffff62a9819cf0d0d9f73e0a5535c0&vid=78ea7a3679ffff62a9819cf0d0d9f73e0a5535c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR