Propylene carbonate ≥98.0%

Supplier: TCI America
Warning

Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester

P0525-25G P0525-500G
TCP0525-25GEA 22 USD
TCP0525-25G TCP0525-500G
Propylene carbonate ≥98.0%
Propylene carbonate

CAS Number: 108-32-7
MDL Number: MFCD00005385
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 242
Melting point (°C): -49
Flash Point (°C): 132
Specific Gravity (20/20): 1.21


Product Categories: Materials Chemistry, Secondary Battery Research Reagents, Monomers, Bioscience, Monomers for Synthetic Polymers (for DDS)
Other Product Names / Synonyms: 4-Methyl-1,3-dioxolan-2-one, 2-Oxo-4-methyl-1,3-dioxolane

Formula: C₄H₆O₃
Boiling Pt: 242 °C
Melting Pt: –49 °C
Density: 1.21 (20 °C)
Flash Pt: 132 °C
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd4f74a96ae6d9c8ed1a11d6e5993155eefbc650&vid=dd4f74a96ae6d9c8ed1a11d6e5993155eefbc650&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR